Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connective Tissue Growth Factor Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071386MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-138-6MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-138-6MI Supplier MilliporeSigma Supplier No. 071386
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Anti-Connective Tissue Growth Factor Antibody detects level of Connective Tissue Growth Factor and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connective Tissue Growth Factor
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified goat polyclonal in buffer containing Tris-Glycine pH 8.0 with 0.05% sodium azide.
Gene Symbols CTGF; CCN2; HCS24; IGFBP8; NOV2
Host Species Goat
Immunogen Recombinant protein corresponding to the mature chain of human Connective tissue growth factor.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001892.1
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.