Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Connexin 26, Polyclonal,
Supplier: MilliporeSigma™ AB8143
Description

Specifications
Connexin 26 | |
Polyclonal | |
Affinity-purified antibody using peptide-Separose column chromatography. Supplied at 1mg/mL in 100mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide. | |
A 19 amino acid peptide sequence located near the C-terminal, second cytoplasmic domain of mouse Cx26 (Henneman et al. 1992; Nishi et al. 1991), conjugated to KLH as a carrier protein. | |
50 μg | |
Mouse |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
Primary | |
−20°C in undiluted aliquotes. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction