Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 35/36 Mouse anti-Rabbit, Fish, Clone: 9D7.2, MilliporeSigma™

Catalog No. MAB3043MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3043MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3043MI Supplier MilliporeSigma Supplier No. MAB3043
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Connexin 35/36 Antibody, clone 9D7.2 detects level of Connexin 35/36 and has been published and validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 35/36
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9D7.2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.1M Citrate buffer with 50% glycerol and 0.1% Sodium Azide as a preservative.
Host Species Mouse
Immunogen Recombinant Perch Connexin 35
Quantity 100 μg
Primary or Secondary Primary
Target Species Fish, Rabbit
Content And Storage −20°C for up to 12 months in undiluted aliquots.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.