Learn More
Connexin 40 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Connexin 40 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 15mM NaCl containing 0.05% sodium azide. |
| Gene Symbols | GJA5; CX40; Connexin-40; MGC11185; Cx40 |
| Host Species | Rabbit |
| Immunogen | Anti-Connexin 40 is made against a 19 amino acid peptide sequence within the C-terminal cytoplasmic domain of mouse C×40 (2). Peptide sequence is conserved in human and rat Cx40 (100%), chicken Cx42 (77%), human Cx50 (87%), rat Cx50 (81%) and sheep Cx49 ( |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.