Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin-43 Mouse, Unlabeled, Clone: 2, BD

Catalog No. BDB610061 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610061 50 μg
BDB610062 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610061 Supplier BD Biosciences Supplier No. 610061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gap junctions are intercellular protein pores. They enable cell-to-cell communication by allowing passage of ions and other small molecules. The subunits of gap junction channels are assembled from a family of proteins called connexins. Individual connexin molecules join to make hexameric hemichannels termed connexons; these structures dock to connexons on neighboring cells, forming gap junction pores. Connexin-43 is a member of the connexin family possessing four transmembrane regions, with cytoplasmic amino and carboxyl terminals. It undergoes rapid turnover in the cell and its monomers may reside in the ER/Golgi network, forming a reservoir available for assembly upon
degradation of existing connexin-43 channels. In addition, it is believed that phosphorylation of connexin-43 plays a regulatory role both in the assembly of connexons and in gating activity at the gap junction.

Host Species: Mouse
Clone: 2
Isotype: IgG2
Species Reactivity: Rat
Immunogen: Rat Connexin-43 aa. 252-270
Formula Weight [Chemical]: 43kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin-43
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Connexin-43 aa. 252-270
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.