Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 43 Mouse anti-Mouse, Human, Rat, Pig, Canine, Clone: 4E6.2, MilliporeSigma™

Catalog No. MAB3067MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3067MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3067MI Supplier MilliporeSigma Supplier No. MAB3067
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Connexin 43 Antibody, clone 4E6.2 detects level of Connexin 43 and has been published and validated for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 43
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4E6.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 liquid in PBS containing sodium azide.
Gene Accession No. NM_000165.3
Gene Symbols GJA1, CX43, GJAL, Cx43, ODD, DFNB38, ODDD, ODOD, SDTY3, Connexin-43
Host Species Mouse
Immunogen A synthetic peptide corresponding to positions 252-270 of the mouse connexin 43 sequence. This sequence is identical in rat and bovine.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Pig, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.