Learn More
Connexin 43 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Connexin 43 |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.02M phosphate buffer, 0.25M NaCl, with 0.1% sodium azide as a preservative. |
| Gene Symbols | GJA1; GJAL; Cx43; DFNB38; SDTY3; CX43; ODD; ODDD; ODOD; Connexin-43 |
| Host Species | Rabbit |
| Immunogen | Anti-Connexin 43 is made against a 23 amino acid C-terminal peptide sequence within the cytoplasmic domain of mouse C×43 (Beyer, E. and Steinberg, T., 1991, JBC 266: 7971). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.