Learn More
Connexin 43 Rabbit anti-Human, Hamster, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Connexin 43, phospho-specific (Ser368) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Protein A and peptide affinity purified immunoglobulin. Liquid in 10mM sodium HEPES, pH 7.5, 150mM NaCl, 100μg/mL BSA, with 50% glycerol. |
| Gene Symbols | GJA1; GJAL; Cx43; DFNB38; SDTY3; CX43; ODD; ODDD; ODOD; Connexin-43 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Ser368 of human Connexin 43. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.