Learn More
Connexin 43 Rabbit anti-Bovine, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Connexin 43 |
| Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity-purified using peptide-Sepharose column chromatography and supplied in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl containing 0.05% sodium azide. |
| Gene Symbols | GJA1; GJAL; Cx43; DFNB38; SDTY3; CX43; ODD; ODDD; ODOD; Connexin-43 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated synthetic peptide corresponding to amino acids 360-382 within the C-terminus of mouse connexin 43. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.