Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 45 Mouse anti-Human, Rat, Rabbit, Clone: 5B9.2, MilliporeSigma™

Catalog No. MAB3101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3101MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3101MI Supplier MilliporeSigma Supplier No. MAB3101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Connexin 45 Antibody, near C-terminus, cytoplasmimmunocytochemistry, clone 5B9.2 detects level of Connexin 45 and has been published and validated for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 45
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5B9.2
Conjugate Unconjugated
Formulation Liquid at 1mg/mL in 0.02M PB with 0.25MNaCl, pH=7.6 containing 0.1% sodium azide.
Gene Symbols ENSG00000182963, Cx45, Connexin-45
Host Species Mouse
Immunogen The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-367 of human connexin 45.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rabbit, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.