Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

coatomer protein complex, subunit beta 1, Mouse, Clone: 3E10, Abnova™

Catalog No. 89000583 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-583 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-583 Supplier Abnova Corporation Supplier No. H00001315M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant COPB.

This gene encodes a protein subunit of the coatomer complex associated with nonclathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described. [provided by RefSeq

Sequence: TVNTNMVDLNDYLQHILKSTNMKCLTPEKALSGYCGFMAANLYARSIFGEDALANVSIEKPIHQGPDAAVTGHIRIRAKSQGMALSLGDKINLSQKKTSI

Specifications

Antigen coatomer protein complex, subunit beta 1
Applications ELISA
Classification Monoclonal
Clone 3E10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant COPB.
Formulation PBS with no preservative; pH 7.4
Gene COPB1
Gene Accession No. NM_016451
Gene Alias COPB/DKFZp761K102/FLJ10341
Gene Symbols COPB1
Host Species Mouse
Immunogen COPB (NP_057535, 854 a.a. ∼ 953 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 1315
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.