Learn More
Coronavirus Group Antigen Mouse anti-Human, Clone: 542-7D, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Coronavirus Group Antigen |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 542-7D |
| Conjugate | Unconjugated |
| Formulation | Purified ascites. Supplied in PBS buffer pH 7.4. Contains 0.1% sodium azide and carrier protein. |
| Host Species | Mouse |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Human |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.