Learn More
Description
Specifications
Specifications
| Antigen | Cortactin, phospho-specific [Tyr466] |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Antibody is cross-adsorbed to nonphospho-peptide corresponding to the site of phosphorylation and preadsorbed with phosphopeptides to cortactin tyrosine 421 and 482 to remove cross-reactivity then affinity purified using phosphorylated cortactin (Tyr466) |
| Gene Symbols | CTTN; EMS1; Amplaxin; 1110020L01Rik; FLJ34459 |
| Host Species | Rabbit |
| Immunogen | Phosphorylated cortactin (Tyr466) synthetic peptide corresponding to mouse amino acid residues around tyrosine 466 (tyrosine 470 in the human sequence). |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.