Learn More
Description
Specifications
Specifications
| Antigen | Cortactin, phospho-specific (Tyr486) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 1mg/mL in PBS with 0.05% Bronidox. |
| Gene Symbols | CTTN; EMS1; Amplaxin; 1110020L01Rik; FLJ34459 |
| Host Species | Rabbit |
| Immunogen | Rabbit anti-Cortactin (Y486) polyclonal antibody was raised against a peptide corresponding to residues surrounding phosphorylated Y486 of human cortactin. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.