Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corticosterone Sheep anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1297MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1297MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1297MI Supplier MilliporeSigma Supplier No. AB1297
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Anti-Corticosterone Antibody is an antibody against Corticosterone for use in ELISA, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Corticosterone
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Sheep serum. Liquid in 0.1% sodium azide.
Host Species Sheep
Immunogen Corticosterone-3-cmo-urease
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.