Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-COUP-TF1, Polyclonal

Catalog No. ABE1425MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1425MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1425MI Supplier MilliporeSigma™ Supplier No. ABE1425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects COUP-TF1 Clone: in Human, Mouse, Rat samples, and it is validated for Chromatin Immunoprecipitation, Immunofluorescence, Immunohistochemistry, Western Blotting

COUP transcription factor 1 (UniProt Q60632; also known as Avian erythroblastic leukemia viral oncogene homolog-like 3, COUP-TF1, COUP-TF I, COUP transcription factor I, EAR-3, Nuclear receptor subfamily 2 group F member 1, V-erbA-related protein 3) is encoded by the Nr2f1 (also known as Erbal3, Tfcoup1) gene (Gene ID 13865) in murine species. COUP transcription factors1&2 (COUP-TFI, COUP-TFII) are two related nuclear orphan receptors implicated in their roles in guiding migrating neurons towards their target regions during brain development. COUP-TFI and COUP-TFII are expressed in distinct and non-overlapping migrating cells in specific pathways in the basal telencephalon of mouse embryos at a stage when interneuron migration is robust. While COUP-TFI appears to control dorsal and ventral migration in the basal telencephalon, COUP-TFII is more specific in modulating tangential migration into the intermediate zone of the cortex. Coup-TFII is also identified as an activator of Prox1 expression in venous lymphatic endothelial cell (LEC) progenitors during early stages of LEC specification and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COUP-TF1
Applications ChIP Assay, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Gene Accession No. Q60632
Gene Symbols Nr2f1, Erbal3, Tfcoup1
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the N-terminal half of mouse COUP-TF1.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_034281
Test Specificity Specificity of the antiserum was confirmed by immunogen blocking in immunohistochemistry on frozen sections (Tripodi, M., et al. (2004). Development. 131(24):6119-6129).
Target Species Human, Mouse, Rat
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.