Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cox-2 Mouse, Unlabeled, Clone: 33, BD

Catalog No. BDB610203 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610203 50 μg
BDB610204 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610203 Supplier BD Biosciences Supplier No. 610203
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclooxygenase (Cox) is also known as prostaglandin H synthase or PGH synthase (E.C. 1.14.99.1). It catalyzes the conversion of arachidonate to prostaglandin H2 (PGH2), the precursor of PGE2, PGF2α, PGD2, prostacyclin, and thromboxane A2. Cox actually has two different enzymatic activities: a cyclooxygenase that mediates the formation of PGG2 from oxygen and arachidonate and a hydroperoxidase that catalyzes a reduction of PGG2 yielding PGH2. Two Cox genes, Cox-1 and Cox-2, have been isolated in several species. A 4kb mRNA encodes the 604 amino acid Cox-2 protein. The two human Cox isoenzymes are 61% identical in amino acid composition with the active sites being highly conserved. Cox-2 mRNA and protein levels are induced by serum, lipopolysaccharides, growth factors, human chorionic gonadotropin and phorbol testers in various mammalian cell types. It has been shown that interleukin-1α (IL-1α) induces increased levels of Cox-2 mRNA and protein in human endothelial cells. The sustained increase in Cox-2 is apparently due (at least in part) to IL-1α increasing the stability of Cox-2 mRNA. This type of regulatory mechanism may play an important role in chronic inflammatory conditions.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cox-2
Applications Western Blot
Classification Monoclonal
Clone 33
Concentration 250μg/mL
Conjugate Unconjugated
Description PGHS-2, Cyclooxygenase-2
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.