Learn More
COX2 Mouse anti-Human, FITC, Clone: AS66, Invitrogen™
Catalog No. 01670943
Change view
Click to view available options
100 Tests; FITC
Catalog No.
01-670-943 100 Tests; FITC
1 options
Catalog No. 01-670-943 Supplier Qorpak Supplier No. 335793
Add to Cart
Add to Cart
Mouse Monoclonal Antibody
This antibody does not cross-react with human COX-1 in ELISA applications.
Cyclooxygenase (COX), also known as Prostaglandin H2 synthase and Prostaglandin endoperoxide synthase, is a key enzyme in the of conversion arachidonic acid to Prostaglandin H2. Prostaglandin H2 is converted by other enzymes into inflammatory mediators prostaglandin (PG) D2, PGE2, PGF2a, PGI2 and Thromboxane A2. Thus, COX is a key enzyme in the production of inflammatory agents and is the target of intense research and drug discovery activities. There are two enzymes of COX, COX-1 and COX-2. COX-1 is constitutively produced in many (most) cell types. It is important in the gut for the production of prostaglandins, which inhibit gastric secretion. It can be induced in monocytes, macrophages and other cells. It is induced as part of the inflammatory response by IL-1beta and other cytokines. It is induced by growth factors such as EGF and PDGF. Expression is inhibited by glucocorticosteroids such as cortisol and dexamethasone. Lipopolysaccharide in bacterial infections induces COX-2. COX-2 is also found in elevated levels in synoviocytes from rheumatoid arthritis patients. The discomforts of inflammation such as pain and swelling are largely due to the action of prostaglandins produced by COX-2.