Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coxsackievirus A9 Mouse anti-Human, Clone: 450-11G-8A-5E, MilliporeSigma™

Catalog No. MAB947MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB947MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB947MI Supplier MilliporeSigma Supplier No. MAB947
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Coxsackievirus A9 Antibody, clone 450-11G-8A-5E is an antibody against Coxsackievirus A9 for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coxsackievirus A9
Applications Immunofluorescence
Classification Monoclonal
Clone 450-11G-8A-5E
Conjugate Unconjugated
Formulation Ascites
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.