Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coxsackievirus B Blend Mouse anti-Human, Clone: Blend of 6 monoclonals, MilliporeSigma™

Catalog No. MAB9410MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9410MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB9410MI Supplier MilliporeSigma Supplier No. MAB9410
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Coxsackievirus B Blend Antibody, clone Blend of 6s is an antibody against Coxsackievirus B Blend for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coxsackievirus B Blend
Applications Immunofluorescence
Classification Monoclonal
Clone Blend of 6 monoclonals
Conjugate Unconjugated
Formulation Ascites.
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.