Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coxsackievirus B1 Mouse anti-Human, Clone: 334-4A-8F-3F, MilliporeSigma™

Catalog No. MAB944MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB944MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB944MI Supplier MilliporeSigma Supplier No. MAB944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Coxsackievirus B1 Antibody, clone 334-4A-8F-3F is validated for use in IF for the detection of Coxsackievirus B1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coxsackievirus B1
Applications Immunofluorescence
Classification Monoclonal
Clone 334-4A-8F-3F
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.