Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coxsackievirus B6 Mouse anti-Human, Clone: 363-9E-2F-11E, MilliporeSigma™

Catalog No. MAB945MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB945MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB945MI Supplier MilliporeSigma Supplier No. MAB945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Coxsackievirus B6 Antibody, clone 363-9E-2F-11E is an antibody against Coxsackievirus B6 for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coxsackievirus B6
Applications Immunofluorescence
Classification Monoclonal
Clone 363-9E-2F-11E
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M PB with 0.25M NaCl, pH=7.6 with 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.