Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPB1, Mouse, Clone: 4D5, Abnova™

Catalog No. 89021146 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-146 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-146 Supplier Abnova Corporation Supplier No. H00001360M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CPB1.

Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma. [provided by RefSeq

Sequence: SAHHGGEHFEGEKVFRVNVEDENHINIIRELASTTQIDFWKPDSVTQIKPHSTVDFRVKAEDTVTVENVLKQNELQYKVLISNLRNVVEAQFDSRVRATGHSYEKYNKWETIEAWTQQVATENPALISRSVIGTTFEGRAIYLLKVGKAGQNKPAIFMDCGFHAREWISPAFCQWFVREAVRTYGREIQVTELLDKLDFYVLPVLNIDGYIYTWTKSRFWRKTRSTHTGSSCIGTDPNRNFDAGWCEIGASRNPCDETYCGPAAESEKETKALADFIRNKLSSIKAYLTIHSYSQMMIYPYSYAYKLGENNAELNALAKATVKELASLHGTKYTYGPGATTIYPAAGGSDDWAYDQGIRYSFTFELRDTGRYGFLLPESQIRATCEETFLAIKYVASYVLEHLY

Specifications

Antigen CPB1
Applications ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CPB1.
Formulation PBS with no preservative; pH 7.4
Gene CPB1
Gene Accession No. BC015338
Gene Alias DKFZp779K1333/PASP/PCPB
Gene Symbols CPB1
Host Species Mouse
Immunogen CPB1 (AAH15338, 14 a.a. ∼ 417 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1360
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.