Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPI-17 Rabbit anti-Bovine, Human, Mouse, Pig, Rat, Rabbit, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07344
Description
Anti-CPI-17 Antibody is an antibody against CPI-17 for use in western blotting.
Specifications
CPI-17 | |
Polyclonal | |
PBS, 0.02% sodium azide before the addition of glycerol to 10% | |
Rabbit | |
Protein A chromatography | |
Primary | |
Bovine, Human, Mouse, Pig, Rabbit, Rat | |
Purified |
Western Blot | |
Unconjugated | |
PPP1R14A; CPI-17; PPP1INL; 17-KDasprotein; CPI17 | |
Recombinant protein, full length pig CPI-17 | |
200 μg | |
NM_033256 | |
−20°C for 24 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction