Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

copine IV, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016614 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-614 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-614 Supplier Abnova Corporation Supplier No. H00131034B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CPNE4 protein.

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene is one of several genes that encode a calcium-dependent protein containing two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. [provided by RefSeq

Sequence: MKKMSNIYESAANTLGIFNSPCLTKVELRVACKGISDRDALSKPDPCVILKMQSHGQWFEVDRTEVIRTCINPVYSKLFTVDFYFEEVQRLRFEVHDISSNHNGLKEADFLGGMECTLGQIVSQRKLSKSLLKHGNTAGKSSITVIAEELSGNDDYVELAFNARKLDDKDFFSKSDPFLEIFRMNDDATQQLVHRTEVVMNNLSPAWKSFKVSVNSLCSGDPDRRLKCIVWDWDSNGKHDFIGEFTSTFKEMRGAMEGKQVQWECINPKYKAKKKNYKNSGTVILNLCKIHKMHSFLDYIMGGCQIQFTVAIDFTASNGDPRNSCSLHYIHPYQPNEYLKALVAVGEICQDYDSDKMFPAFGFGARIPPEYTVSHDFAINFNEDNPECAGIQGVVEAYQSCLPKLQLYGPTNIAPIIQKVAKSASEETNTKEASQYFILLILTDGVITDMADTREAIVHASHLPMSVIIVGVGNADFSDMQMLDGDDGILRSPKGEPVLRDIVQFVPFRNFKHASPAALAKSVLAEVPNQVVDYYNGKGIKPKCSSEMYESSRTLAP

Specifications

Antigen copine IV
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CPNE4 protein.
Formulation PBS with no preservative; pH 7.4
Gene CPNE4
Gene Accession No. NM_130808.1
Gene Alias COPN4/CPN4/MGC15604
Gene Symbols CPNE4
Host Species Mouse
Immunogen CPNE4 (NP_570720.1, 1 a.a. ∼ 557 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131034
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.