Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT1A Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS65
Description
Detect CPT1A using this Anti-CPT1A Antibody validated for use in Western Blotting.
Specifications
CPT1A | |
Polyclonal | |
Rabbit polyclonal serum containing 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Mouse, Rat | |
Serum |
Western Blot | |
Unconjugated | |
CPT1A; CPT1 | |
GST-tagged recombinant protein corresponding to rat CPT1A. | |
100 μL | |
NP_001867 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction