Learn More
carnitine palmitoyltransferase II, Mouse, Clone: 1G7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CPT2.
Supplier: Abnova Corporation H00001376M02
Description
The protein encoded by this gene is a nuclear protein which is transported to the mitochondrial inner membrane. Together with carnitine palmitoyltransferase I, the encoded protein oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders. [provided by RefSeq
Sequence: WFDKSFNLIIAKDGSTAVHFEHSWGDGVAVLRFFNEVFKDSTQTPAVTPQSQPATTDSTVTVQKLNFELTDALKTGITAAKEKFDATMKTLTIDCVQFQRSpecifications
carnitine palmitoyltransferase II | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC005172 | |
CPT2 | |
CPT2 (AAH05172, 351 a.a. ∼ 450 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
1G7 | |
Mouse monoclonal antibody raised against a partial recombinant CPT2. | |
CPT2 | |
CPT1/CPTASE | |
Mouse | |
Affinity Purified | |
RUO | |
1376 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.