Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRABP2 Mouse anti-Human, Mouse, MilliporeSigma™

Catalog No. MAB5488MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5488MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5488MI Supplier MilliporeSigma Supplier No. MAB5488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-CRABP2 Antibody (Mouse Monoclonal Antibody) validated in ELISA, western blot, immunocytochemistry to detect CRABP2 also known as Cellular Retinoic Acid Binding Protein II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRABP2
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. NM_001878.2
Gene Symbols CRABP2, CRABP-II, RBP6, CRABPII
Host Species Mouse
Immunogen Full length recombinant human CRABPII.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.