Learn More
CRABP2 Mouse anti-Human, Unlabeled, Clone: M60-1228, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560234
Description
The M60-1228 monoclonal antibody reacts with Cellular Retinoic Acid-Binding Protein 2 (CRABP2). CRABP2 is a cytosolic low-molecular-weight protein with high affinity for retinoic acid (RA), a member of the vitamin A family that is able to induce cell differentiation. The translocation of CRABP2 to the nucleus indicates that it may function to deliver RA to RA receptors, which activate gene transcription. The differential expression of the CRABP2 gene during development and its overexpression in many cancers suggest that it is important in RA-mediated regulation of cell growth and differentiation.
Host Species: Mouse
Clone: M60-1228
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human CRABP2 Peptide

Specifications
CRABP2 | |
Monoclonal | |
0.5mg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG1 |
In vitro Assay, Western Blot | |
M60-1228 | |
Unconjugated | |
Human CRABP2 Peptide | |
0.1 mg | |
Cell Biology | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.