Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cranin Mouse anti-Chicken, Human, Mouse, Rat, Sheep, Xenopus, Clone: 6C1, MilliporeSigma™

Catalog No. MAB371MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB371MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB371MI Supplier MilliporeSigma Supplier No. MAB371
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cranin Antibody, aa572-604 of human sequence, clone 6C1 is validated for use in ELISA, immunocytochemistry, immunohistochemistry for the detection of Cranin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cranin
Applications ELISA, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 6C1
Conjugate Unconjugated
Formulation Ascites Liquid.
Host Species Mouse
Immunogen Synthetic peptide corresponding to a.a. 572-604 of human cranin.
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat, Sheep, Xenopus species
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.