Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cre Recombinase Mouse anti-All, Clone: 2D8, MilliporeSigma™

Catalog No. MAB3120MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3120MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3120MI Supplier MilliporeSigma Supplier No. MAB3120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cre Recombinase Antibody, clone 2D8 is a well characterized monoclonal antibody. Proven to reliably detect Cre Recombinase, this mAb is validated for use in ELISA, immunocytochemistry, immunofluoresence, immunohistochemistry and western blotting and is backed by multiple publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cre Recombinase
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2D8
Conjugate Unconjugated
Formulation Ascites fluid. Contains no preservative
Host Species Mouse
Immunogen Cre recombinase (aa 77 to 343) fusion protein
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C for up to 12 months.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.