Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06863MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-863-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-863-MI Supplier MilliporeSigma Supplier No. 06863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CREB Antibody, Cat. No. 06-863, is a highly specific rabbit Polyclonal antibody, that targets CREB and has been tested in Chromatin Immunoprecipitation (ChIP), ChIP-seq, Electrophoretic Mobility Shift Assay (EMSA), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, and 30% glycerol.
Gene Symbols MGC9284; CREB1
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (CSGAENQ QSGDAAVTEAENQQ) corresponding to a.a. 5-24 of human CREB. The immunizing sequence is identical in pig, shares 19 of 20 residues with mouse and bovine and 18 of 20 residues with rat.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_134442
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.