Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBP, Polyclonal, Invitrogen™

Catalog No. 01673187
Change view
Click to view available options
Quantity:
7 mL
Catalog No. Quantity
01-673-187 7 mL
1 options

Catalog No. 01-673-187

Supplier: Thermo Scientific PA137264

Rabbit Polyclonal Antibody

CBP for CREB-binding protein is a 265kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene CREBBP
Gene Accession No. Q92793
Gene Alias CREB Binding Protein
Gene Symbols CREBBP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human CBP
Purification Method Antigen affinity chromatography
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1387
Target Species Human
Content And Storage Store at 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.