Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB Mouse anti-Human, Mouse, Rat, MilliporeSigma™

Catalog No. MAB5432MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5432MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5432MI Supplier MilliporeSigma Supplier No. MAB5432
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CREB Antibody is a Mouse Monoclonal Antibody for detection of CREB also known as cAMP-response Element Binding Protein and has been validated in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.
Gene Accession No. NM_004379.2; NM_134442.2
Gene Symbols CREB1, MGC9284, CREB
Host Species Mouse
Immunogen Recombinant CREB-A protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.