Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB (pS133) / ATF-1 (pS63) Mouse, Alexa Fluor 647, Clone: J151-21, BD

Catalog No. BDB558434 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558434 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558434 Supplier BD Biosciences Supplier No. 558434
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcription of various genes is regulated by the cyclic AMP (cAMP) signal transduction pathway through a family of cAMP Responsive Element (CRE)-Binding transcription factors that include CREB, CREM (CRE Modulator), and ATF-1 (Activating Transcription Factor 1). The genes for these transcription factors encode multiple isoforms that are created by alternative splicing, alternative initiation codons, and alternative intronic promoters. Homo- and heterodimers of these members of the basic domain-and-leucine zipper superfamily of proteins bind the palindromic TGACGTCA sequence of CRE. CREB is expressed in all somatic cells, and many different stimuli activate CREB by phosphorylation of its serine 133 (S133). The transcriptional activity and specificity of CREB are regulated by the signaling pathways initiated by the various stimuli. Transcriptional regulation by CREB family members has been implicated in many physiological responses to extracellular and environmental stimuli, such as memory, tissue growth and development, and homeostasis. The J151-21 monoclonal antibody recognizes the phosphorylated S133 of human CREB in addition to cross-reacting with S63 of human ATF-1.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB (pS133) / ATF-1 (pS63)
Applications Flow Cytometry
Classification Monoclonal
Clone J151-21
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human CREB Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.