Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB Rabbit anti-Human, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3442MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3442MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3442MI Supplier MilliporeSigma Supplier No. AB3442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CREB Antibody, pSer133 is a Rabbit Polyclonal Antibody for detection of CREB also known as CREB-1, cAMP-response Element-binding Protein-1 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB, pSer133
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid. The antibody has been preabsorbed with unphosphorylated peptide. Contains 0.01% sodium azide.
Gene Symbols CREB1; MGC9284; CREB
Host Species Rabbit
Immunogen Phosphopeptide conjugated to KLH.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004379.2; NM_134442.2
Target Species Human, Monkey, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.