Learn More
cAMP responsive element binding protein 5, Mouse, Clone: 8A5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CREB5.
Supplier: Abnova Corporation H00009586M02
Description
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MFCTSGGNSASVMSMRPVPGSLSSLLHLHNRQRQPMPASMPGTLPNPTMPGSSAVLMPMERQMSVNSSIMGMQGPNLSNPCASPQVQPMHSEAKMRLKASpecifications
cAMP responsive element binding protein 5 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001011666 | |
CREB5 | |
CREB5 (NP_001011666, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human, Mouse | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
8A5 | |
Mouse monoclonal antibody raised against a partial recombinant CREB5. | |
CREB5 | |
CRE-BPA | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
9586 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.