Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cAMP responsive element binding protein-like 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005171 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-171 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-171 Supplier Abnova Corporation Supplier No. H00001389B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CREBL2 protein.

cAMP response element (CRE)-binding protein-like-2 (CREBL2) was identified in a search to find genes in a commonly deleted region on chromosome 12p13 flanked by ETV6 and CDKN1B genes, frequently associated with hematopoietic malignancies, as well as breast, nonsmall-cell lung and ovarian cancers. CREBL2 shares a 41% identity with CRE-binding protein (CREB) over a 48-base long region which encodes the bZip domain of CREB. The bZip domain consists of about 30 amino acids rich in basic residues involved in DNA binding, followed by a leucine zipper motif involved in protein dimerization. This suggests that CREBL2 encodes a protein with DNA binding capabilities. The occurance of CREBL2 deletion in malignancy suggests that CREBL2 may act as a tumor suppressor gene. [provided by RefSeq

Sequence: MDDSKVVGGKVKKPGKRGRKPAKIDLKAKLERSRQSARECRARKKLRYQYLEELVSSRERAICALREELEMYKQWCMAMDQGKIPSEIKALLTGEEQNKSQQNSSRHTKAGKTDANSNSW

Specifications

Antigen cAMP responsive element binding protein-like 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CREBL2 protein.
Formulation No additive
Gene CREBL2
Gene Accession No. NM_001310.2
Gene Alias MGC117311/MGC138362
Gene Symbols CREBL2
Host Species Mouse
Immunogen CREBL2 (NP_001301.1, 1 a.a. ∼ 120 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 1389
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.