Learn More
cysteine-rich with EGF-like domains 2, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | cysteine-rich with EGF-like domains 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant CRELD2. |
| Formulation | 50% glycerol |
| Gene | CRELD2 |
| Gene Accession No. | NM_024324 |
| Gene Alias | DKFZp667O055/MGC11256 |
| Gene Symbols | CRELD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.