Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216009
Description
CREM Polyclonal specifically detects CREM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CREM | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| CREM | |
| Recombinant protein encompassing a sequence within the center region of human CREM. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 1390 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.31mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| cAMP response element modulator, cAMP responsive element modulator, cAMP-responsive element modulator, hCREM 2alpha-b protein, hCREM 2beta-a protein, hCREM-2, ICER, Inducible cAMP early repressor, inducible cAMP early repressor ICER, MGC111110, MGC17881, MGC41893 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction