Learn More
corticotropin releasing hormone, Mouse, Clone: 2B11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CRH.
Supplier: Abnova Corporation H00001392M02
Description
Corticotropin-releasing hormone (CRH) is a 41-amino acid peptide derived from a 191-amino acid preprohormone. CRH is secreted by the paraventricular nucleus (PVN) of the hypothalamus in response to stress. Marked reduction in CRH has been observed in association with Alzheimer disease and autosomal recessive hypothalamic corticotropin dificiency has multiple and potentially fatal metabolic consequences including hypoglycemia and hepatitis. In addition to production in the hypothalamus, CRH is also synthesized in peripheral tissues, such as T lymphocytes and is highly expressed in the placenta. In the placenta CRH is a marker that determines the length of gestation and the timing of parturition and delivery. A rapid increase in circulating levels of CRH occurs at the onset of parturition, suggesting that, in addition to its metabolic functions, CRH may act as a trigger for parturition. [provided by RefSeq
Sequence: SEEPPISLDLTFHLLREVLEMARAEQLAQQAHSNRKLMEIIGKSpecifications
corticotropin releasing hormone | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011031 | |
CRH | |
CRH (AAH11031, 154 a.a. ∼ 196 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
1392 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Western Blot | |
2B11 | |
Mouse monoclonal antibody raised against a partial recombinant CRH. | |
CRH | |
CRF | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.