Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRIPT, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127799
Description
PA1-27799 detects CRIPT from human, mouse, and rat samples.PA1-27799 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27679 immunogen is a synthetic peptide corresponding to human HSPC139.
Specifications
CRIPT | |
Polyclonal | |
CRIPT | |
HSPC139 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
9419 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q9P021, O70333, Q792Q4 | |
CRIPT | |
Synthetic peptide corresponding to human HSPC139. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction