Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CrkL (pY207) Mouse, Alexa Fluor 488, Clone: K30-391.50.80, BD

Catalog No. BDB560789 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560789 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560789 Supplier BD Biosciences Supplier No. 560789

Mouse Monoclonal Antibody

Crk-Like (CrkL) is an adaptor protein that is preferentially expressed in hematopoietic cells and is encoded by a gene that is homologous to the viral oncogene v-crk (cegulator of kinase). Its SH2 and SH3 domains bind to a variety of effector proteins, such as paxillin, p130Cas, c-Cbl, c-Abl, and C3G. These interactions are involved in the regulation of cellular migration, adhesion, and transformation. Tyrosine 207 (Y207) of CrkL is phosphorylated in hematopoietic cells that express the BCR-ABL fusion protein. This site may be a negative regulator of protein complex formation and biological activity.

The K30-391.50.80 monoclonal antibody recognizes the phosphorylated Y207 of human CrkL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CrkL (pY207)
Applications Flow Cytometry
Classification Monoclonal
Clone K30-391.50.80
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human, Mouse, and Rat CrkL Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.