Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mediator complex subunit 7, Mouse, Clone: 2D7, Abnova™

Catalog No. 89001135 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-135 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-135 Supplier Abnova Corporation Supplier No. H00009443M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CRSP9.

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: MGEPQQVSALPPPPMQYIKEYTDENIQEGLAPKPPPPIKDSYMMFGNQFQCDDLIIRPLESQGIERLHPMQFDHKKELRKLNMSILINFLDLLDILIRSPGSIKREEKLEDLKLLFVHVHHLINEYRPHQARETLRVMMEVQKRQRLETAERFQKHLERVIEMIQNCLASLPDDLPHSEAGMRVKTEPMDADDSNNCTGQNEHQRENSGHRRDQIIEKDAALCVLIDEMNERP

Specifications

Antigen mediator complex subunit 7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CRSP9.
Formulation PBS with no preservative; pH 7.4
Gene MED7
Gene Accession No. BC005250
Gene Alias CRSP33/CRSP9/MGC12284
Gene Symbols MED7
Host Species Mouse
Immunogen CRSP9 (AAH05250, 1 a.a. ∼ 233 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9443
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.