Learn More
anti-CRTH-2, Alexa Fluor 488, Clone: 301109, R&D Systems™
Chemoattractant Receptor-homologous Molecule Expressed on Th2 Cells
Supplier: R&D Systems FAB33381G
Description
CRTH-2 is a 7-transmembrane G-protein coupled receptor that is expressed by activated Th2 cells and eosinophils. CRTH-2 binds prostaglandin D2 and induces chemotaxis of Th2 cells and eosinophils in allergic and inflammatory immune responses. Human and mouse CRTH-2 share 78% amino acid sequence identity.
Host Species: Mouse
Clone: 301109
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CRTH-2. Met1-Ser395; Accession Number Q9Y5Y4
Applications:
Flow Cytometry
Order Info
The product is shipped with polar packs.
Specifications
CRTH-2/GPR44 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, GPR44, PGD2R, PTGDR2, putative G-protein coupled receptor 44 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11251 | |
Human | |
Purified |
Flow Cytometry | |
301109 | |
Flow Cytometry 5 uL/10^6 cells | |
Q9Y5Y4 | |
PTGDR2 | |
NS0 mouse myeloma cell line transfected with human CRTH-2, Met1-Ser395, Accession # Q9Y5Y4 | |
100 Tests | |
Primary | |
Detects human CRTH-2 in Cell-Based ELISA. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.