Learn More
cone-rod homeobox, Mouse, Clone: F6-C2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CRX.
Supplier: Abnova Corporation H00001406M01
Description
The protein encoded by this gene is a photoreceptor-specific transcription factor which plays a role in the differentiation of photoreceptor cells. This homeodomain protein is necessary for the maintenance of normal cone and rod function. Mutations in this gene are associated with photoreceptor degeneration, Leber congenital amaurosis type III and the autosomal dominant cone-rod dystrophy 2. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined. [provided by RefSeq
Sequence: MMAYMNPGPHYSVNALALSGPSVDLMHQAVPYPSAPRKQRRERTTFTRSQLEELEALFAKTQYPDVYAREEVALKINLPESRVQVWFKNRRAKCRQQRQQQKQQQQPPGGQAKARPAKRKAGTSPRPSTDVCPDPLGISDSYSPPLPGPSGSPTTAVATVSIWSPASESPLPEAQRAGLVASGPSLTSAPYAMTYAPASAFCSSPSAYGSPSSYFSGLDPYLSPMVPQLGGPALSPLSGPSVGPSLAQSPTSLSGQSYGAYSPVDSLEFKDPTGTWKFTYNPMDPLDYKDQSAWKFQILSpecifications
cone-rod homeobox | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC016664 | |
CRX | |
CRX (AAH16664, 1 a.a. ∼ 300 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, KnockDown, Western Blot | |
F6-C2 | |
Mouse monoclonal antibody raised against a full length recombinant CRX. | |
CRX | |
CORD2/CRD/LCA7/OTX3 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
1406 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.