Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CRYAB, Clone: 4E8-1C5, Abnova™

Catalog No. 89021170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-170 Supplier Abnova Corporation Supplier No. H00001410M02
Add to Cart
Add to Cart

Mouse Monoclonal

Host Species: Mouse
Clone: 4E8-1C5
Isotype: IgG1, κ
Cross Reactivity: Human
Immunogen: CRYAB (AAH07008, 1 a.a. ∽176 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26kDa.

ELISA, Sandwich ELISA, Western Blotting

Specifications

Antigen CRYAB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E8-1C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CRYAB.
Formulation PBS with no preservative; pH 7.4
Gene CRYAB
Gene Accession No. BC007008
Gene Alias CRYA2/CTPP2/HSPB5
Gene Symbols CRYAB
Host Species Mouse
Immunogen CRYAB (AAH07008, 1 a.a. ∼ 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1410
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.