Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CRYAB, Monoclonal, Abnova™

Catalog No. 89208063 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-208-063 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-208-063 Supplier Abnova Corporation Supplier No. H00001410K

Host Species: Rabbit
Isotype:IgG
Species Reactivity:Hu
Immunogen:A synthetic peptide of human CRYAB

Customer may provide cell or tissue lysate for antibody screening.
Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Enzyme Linked Immunosorbent Assay, Western Blotting

Order Info

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Specifications

Antigen CRYAB
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description crystallin, alpha B
Formulation In 1X PBS, pH 7.4
Gene CRYAB
Gene Accession No. CRYAB
Gene Alias CRYA2, CTPP2, HSPB5
Gene Symbols CRYAB
Host Species Rabbit
Immunogen A synthetic peptide of human CRYAB
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1410
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.