Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYBA2, Mouse, Clone: 2G9, Abnova™

Catalog No. 89021171 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-171 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-171 Supplier Abnova Corporation Supplier No. H00001412M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CRYBA2.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of the vertebrate eye, which function to maintain the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also defined as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group but absent in the acidic group). Beta-crystallins form aggregates of different sizes and are able to form homodimers through self-association or heterodimers with other beta-crystallins. This gene is a beta acidic group member. Three alternatively spliced transcript variants encoding identical proteins have been reported. [provided by RefSeq

Sequence: SFRPVLCANHNDSRVTLFEGDNFQGCKFDLVDDYPSLPSMGWASKDVGSLKVSSGAWVAYQYPGYRGYQYVLERDRHSGEFCTYGELGTQAHTGQLQSIR

Specifications

Antigen CRYBA2
Applications ELISA
Classification Monoclonal
Clone 2G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CRYBA2.
Formulation PBS with no preservative; pH 7.4
Gene CRYBA2
Gene Accession No. NM_005209
Gene Symbols CRYBA2
Host Species Mouse
Immunogen CRYBA2 (NP_005200.1, 94 a.a. ∼ 193 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1412
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.