Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF1 Mouse anti-Human, Clone: 2D10, MilliporeSigma™

Catalog No. MABF191MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF191MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF191MI Supplier MilliporeSigma Supplier No. MABF191
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CSF1 Antibody, clone 2D10 is a highly specific mouse monoclonal antibody, that targets CSF1 and has been tested in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSF1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Gene Symbols CSF1
Host Species Mouse
Immunogen Purified recombinant fragment of human CSF1 expressed in E. Coli.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.